CH6371376843 Chem Impex International, Inc.

5-(4-Nitrophenyl)-1H-tetrazole

  • MDL number MFCD00068729
  • Cas Number : 16687-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-226 �C
Purity 98
Storage temp. Store at RT
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-4-nitrophenyl-1h-tetrazole/667?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-4-nitrophenyl-1h-tetrazole/667?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart