CH6371376852 Chem Impex International, Inc.

N-(2-Acetamido)-2-iminodiacetic acid monosodium salt

  • MDL number MFCD00065483
  • Cas Number : 7415-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym ADA
Purity 99
Storage temp. Store at RT
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-2-acetamido-2-iminodiacetic-acid-monosodium-salt/692?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-2-acetamido-2-iminodiacetic-acid-monosodium-salt/692?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart