CH6371376942 Chem Impex International, Inc.

Thymol blue sodium salt

  • MDL number MFCD00001617
  • Cas Number : 81012-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Thymolsulfonephthalein, sodium salt
Storage temp. Store at RT
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/thymol-blue-sodium-salt/788?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/thymol-blue-sodium-salt/788?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart