CH6371376964 Chem Impex International, Inc.

L-2,3-Diaminopropionic acid hydrochloride

  • MDL number MFCD00065497
  • Cas Number : 1482-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Dap-OH HCl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-3-diaminopropionic-acid-hydrochloride/814?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-3-diaminopropionic-acid-hydrochloride/814?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart