CH6371377006 Chem Impex International, Inc.

Boc-D-leucine hydrate

  • MDL number MFCD00065583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-89 �C
Synonym Boc-D-Leu-OH H2O
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-leucine-hydrate/858?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-leucine-hydrate/858?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart