CH6371377030 Chem Impex International, Inc.

Nalpha-Boc-Nin-mesitylene-2-sulfonyl-L-tryptophan dicyclohexylammonium salt

  • MDL number MFCD00065596
  • Cas Number : 92916-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-182 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-in-sup-mesitylene-2-sulfonyl-l-tryptophan-dicyclohexylammonium-salt/882?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-in-sup-mesitylene-2-sulfonyl-l-tryptophan-dicyclohexylammonium-salt/882?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart