CH6371377036 Chem Impex International, Inc.

Boc-O-2,6-dichlorobenzyl-D-tyrosine

  • MDL number MFCD00037107
  • Cas Number : 69541-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Tyr(2,6-di-Cl-Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-d-tyrosine/888?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-d-tyrosine/888?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart