CH6371377061 Chem Impex International, Inc.

Ala-Glu-OH

  • MDL number MFCD00025556
  • Cas Number : 13187-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Alanyl-L-glutamic acid
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-glu-oh/914?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-glu-oh/914?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart