CH6371377094 Chem Impex International, Inc.

N6-Benzyladenosine

  • MDL number MFCD00005740
  • Cas Number : 4294-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Benzylaminopurine riboside
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/n-sup-6-sup-benzyladenosine/947?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/n-sup-6-sup-benzyladenosine/947?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart