CH6371377160 Chem Impex International, Inc.

4-Methylumbelliferyl-N-acetyl-b-D-galactosaminide

  • MDL number MFCD00067658
  • Cas Number : 36476-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Methylumbelliferyl-2-acetamido-2-deoxy-beta-O-galactopyranoside
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-n-acetyl-ß-d-galactosaminide/1021?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methylumbelliferyl-n-acetyl-ß-d-galactosaminide/1021?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart