CH6371377209 Chem Impex International, Inc.

Nalpha-Boc-Ndelta-2-chloro-Z-L-ornithine

  • MDL number MFCD00076972
  • Cas Number : 118554-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-118° C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-auml-sup-2-chloro-z-l-ornithine/1075?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-auml-sup-2-chloro-z-l-ornithine/1075?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart