CH6371377213 Chem Impex International, Inc.

Sodium bisulfite

  • MDL number MFCD00003530
  • Cas Number : 7631-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 58.5
Density 1.48
Storage temp. Store at RT
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/sodium-bisulfite/1079?cid=0

Safety Information

hcodes 9
MSDS http://www.chemimpex.com/product/productinfo/sodium-bisulfite/1079?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart