CH6371377217 Chem Impex International, Inc.

Nalpha-Z-Nepsilon-Boc-L-lysine-N-hydroxysuccinimide ester

  • MDL number MFCD00038822
  • Cas Number : 3338-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-97 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White Powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-boc-l-lysine-i-n-i-hydroxysuccinimide-ester/1084?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-z-i-n-i-sup-e-sup-boc-l-lysine-i-n-i-hydroxysuccinimide-ester/1084?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart