CH6371377237 Chem Impex International, Inc.

Basic fuchsin Certified

  • MDL number MFCD00012569
  • Cas Number : 632-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248-252� C
Synonym Basic violet 14
Storage temp. Store at RT
Physical Form Dark Green Solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/basic-fuchsin-certified/1107?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/basic-fuchsin-certified/1107?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart