CH6371377274 Chem Impex International, Inc.

Rhodamine B isothiocyanate - BioReagent, mixed isomers, suitable for protein labelling.

  • MDL number MFCD00041787
  • Cas Number : 36877-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at = -20 °C
Physical Form Red to purple granular powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/rhodamine-b-isothiocyanate-mixture-of-isomers/1152?cid=0

Safety Information

hcodes 9
MSDS http://www.chemimpex.com/product/productinfo/rhodamine-b-isothiocyanate-mixture-of-isomers/1152?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart