CH6371377289 Chem Impex International, Inc.

Acetoacetyl-L-carnitine chloride

  • MDL number MFCD00274126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetoacetyl-l-carnitine-chloride/1187?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetoacetyl-l-carnitine-chloride/1187?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart