CH6371377293 Chem Impex International, Inc.

Decanoyl-L-carnitine chloride

  • MDL number MFCD00274129
  • Cas Number : 14919-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Capryl-L-carnitine chloride
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/decanoyl-l-carnitine-chloride/1191?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/decanoyl-l-carnitine-chloride/1191?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart