CH6371377296 Chem Impex International, Inc.

Linoleoyl-L-carnitine chloride

  • MDL number MFCD00274132

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at =-4°C
Physical Form Viscous solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/linoleoyl-l-carnitine-chloride/1194?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/linoleoyl-l-carnitine-chloride/1194?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart