CH6371377302 Chem Impex International, Inc.

Propionyl-L-carnitine chloride

  • MDL number MFCD00274137
  • Cas Number : 119793-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-159 �C
Purity 98
Storage temp. Store at = -4
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/propionyl-l-carnitine-chloride/1200?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/propionyl-l-carnitine-chloride/1200?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart