CH6371377305 Chem Impex International, Inc.

D-Carnitine hydrochloride

  • MDL number MFCD00064131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Store at RT
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-carnitine-hydrochloride/1203?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-carnitine-hydrochloride/1203?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart