CH6371377311 Chem Impex International, Inc.

Myristoyl coenzyme A monohydrate

  • MDL number MFCD00077829
  • Cas Number : 3130-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym n-Tetradecanoyl coenzyme A monohydrate
Purity 0.94
Storage temp. Store at = -15
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/myristoyl-coenzyme-a/1209?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/myristoyl-coenzyme-a/1209?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart