CH6371377313 Chem Impex International, Inc.

Octanoyl coenzyme A

  • MDL number MFCD02259055
  • Cas Number : 1264-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at = -15 °C
Physical Form White amorphous powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/octanoyl-coenzyme-a/1211?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/octanoyl-coenzyme-a/1211?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart