CH6371377321 Chem Impex International, Inc.

2-Cyanoethylphosphorodichloridite

  • MDL number MFCD00040958
  • Cas Number : 76101-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2-Cyanoethoxy)dichlorophosphine
Purity 8831.
Storage temp. Store at =-15�Keep under Argon or Nitrogen.
Physical Form Colorless to pale yellow liquid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Boiling point 105-108 �2 mm
Product Link http://www.chemimpex.com/product/productinfo/2-cyanoethylphosphorodichloridite/1237?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-cyanoethylphosphorodichloridite/1237?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart