CH6371377370 Chem Impex International, Inc.

Phosphorus pentoxide

  • MDL number MFCD00011440
  • Cas Number : 1314-56-3
  • Molecular Weight : 141.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 350 - 360 °C
Purity 99.50
Density 2.3
Storage temp. Store at RT
Physical Form White soft powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phosphorus-pentoxide/1312?cid=0

Safety Information

hcodes 8
MSDS http://www.chemimpex.com/product/productinfo/phosphorus-pentoxide/1312?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart