CH6371377380 Chem Impex International, Inc.

2,6-Diaminopurine-2'-deoxyriboside

  • MDL number MFCD00047240
  • Cas Number : 4546-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 9-(2-Deoxy-beta-D-ribofuranosyl)-2,6-diaminopurine
Purity 0.99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-6-diaminopurine-2-deoxyriboside/1323?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-6-diaminopurine-2-deoxyriboside/1323?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart