CH6371377390 Chem Impex International, Inc.

Fmoc-S-acetamidomethyl-L-penicillamine

  • MDL number MFCD00151935
  • Cas Number : 201531-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Pen(Acm)-OH
Purity 96
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-penicillamine/1334?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-acetamidomethyl-l-penicillamine/1334?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart