CH6371377399 Chem Impex International, Inc.

3'-O-Acetylthymidine

  • MDL number MFCD00056059
  • Cas Number : 21090-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at =-15°C
Physical Form White to off-white free flowing powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-o-i-acetylthymidine/1344?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-o-i-acetylthymidine/1344?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart