CH6371377417 Chem Impex International, Inc.

Z-D-homophenylalanine

  • MDL number MFCD00066107
  • Cas Number : 138812-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-108° C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-homophenylalanine/1363?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-homophenylalanine/1363?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart