CH6371377421 Chem Impex International, Inc.

Na-Z-D-arginine

  • MDL number MFCD00063009
  • Cas Number : 6382-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-97� C
Synonym Z-D-Arg-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-arginine/1367?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-d-arginine/1367?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart