CH6371377423 Chem Impex International, Inc.

Na-Z-L-arginine-4-nitroanilide hydrochloride

  • MDL number MFCD00058247
  • Cas Number : 59188-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-182
Synonym Z-L-Arg-pNA HCl
Purity 99
Storage temp. Store at 0-8
Physical Form Yellowish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-arginine-4-nitroanilide-hydrochloride/1371?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-arginine-4-nitroanilide-hydrochloride/1371?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart