CH6371377424 Chem Impex International, Inc.

Na-Z-Nw-(4-toluenesulfonyl)-L-arginine

  • MDL number MFCD00077010
  • Cas Number : 13650-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-83
Synonym Z-L-Arg(Tos)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-4-toluenesulfonyl-l-arginine/1372?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-4-toluenesulfonyl-l-arginine/1372?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart