CH6371377427 Chem Impex International, Inc.

Nalpha-Z-L-asparagine 2-nitrophenyl ester

  • MDL number MFCD00274142
  • Molecular Weight : 429.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 162 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-2-nitrophenyl-ester/1375?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-z-l-asparagine-2-nitrophenyl-ester/1375?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart