CH6371377428 Chem Impex International, Inc.

Na-Z-L-asparagine 4-nitrophenyl ester

  • MDL number MFCD00024661
  • Cas Number : 3256-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Asn-ONp
Purity 0.98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-asparagine-4-nitrophenyl-ester/1376?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-l-asparagine-4-nitrophenyl-ester/1376?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart