CH6371377432 Chem Impex International, Inc.

Z-L-aspartic acid beta-benzyl ester alpha-4-nitrophenyl ester

  • MDL number MFCD00057618
  • Cas Number : 2419-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-78 °C
Purity 97
Storage temp. Store at 0-8 °C
Physical Form Cream powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-beta-benzyl-ester-alpha-4-nitrophenyl-ester/1381?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-beta-benzyl-ester-alpha-4-nitrophenyl-ester/1381?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart