CH6371377436 Chem Impex International, Inc.

Z-Glu-Phe-OH

  • MDL number MFCD00037262
  • Cas Number : 987-84-8
  • Molecular Weight : 428.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-169 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-glu-phe-oh/1385?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-glu-phe-oh/1385?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart