CH6371377442 Chem Impex International, Inc.

Z-D-glutamic acid

  • MDL number MFCD00063193
  • Cas Number : 63648-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-D-Glu-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-glutamic-acid/1392?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-glutamic-acid/1392?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart