CH6371377449 Chem Impex International, Inc.

Z-glycine 4-nitrophenyl ester

  • MDL number MFCD00024663
  • Cas Number : 1738-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-132 �C
Synonym Z-Gly-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-glycine-4-nitrophenyl-ester/1399?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-glycine-4-nitrophenyl-ester/1399?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart