CH6371377452 Chem Impex International, Inc.

Z-Gly-Gly-Gly-Gly-Gly-OH

  • MDL number MFCD00077019
  • Cas Number : 20228-72-2
  • Molecular Weight : 437.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-gly-gly-gly-gly-gly-oh/1402?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-gly-gly-gly-gly-gly-oh/1402?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart