CH6371377456 Chem Impex International, Inc.

Z-His-NH NH2

  • MDL number MFCD00038621
  • Cas Number : 49706-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-170 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder with clumps
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-his-nh-font-size-2-face-symbol-middot-font-nh-sub-2-sub/1406?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-his-nh-font-size-2-face-symbol-middot-font-nh-sub-2-sub/1406?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart