CH6371377461 Chem Impex International, Inc.

Z-L-isoleucine-N-hydroxysuccinimide ester

  • MDL number MFCD00079002
  • Cas Number : 3391-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-Ile-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-isoleucine-i-n-i-hydroxysuccinimide-ester/1412?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-isoleucine-i-n-i-hydroxysuccinimide-ester/1412?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart