CH6371377466 Chem Impex International, Inc.

Z-L-leucine-N-hydroxysuccinimide ester

  • MDL number MFCD00042759
  • Cas Number : 3397-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112-119
Synonym Z-L-Leu-OSu
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-leucine-i-n-i-hydroxysuccinimide-ester/1417?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-leucine-i-n-i-hydroxysuccinimide-ester/1417?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart