CH6371377472 Chem Impex International, Inc.

Nalpha-Z-L-lysine benzyl ester benzenesulfonate salt

  • MDL number MFCD00077024
  • Cas Number : 68973-36-4
  • Molecular Weight : 528.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-l-lysine-benzyl-ester-benzenesulfonate-salt/1423?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-l-lysine-benzyl-ester-benzenesulfonate-salt/1423?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart