CH6371377479 Chem Impex International, Inc.

Nepsilon-Z-D-lysine methyl ester hydrochloride

  • MDL number MFCD00077026
  • Cas Number : 145586-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116 - 120 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-d-lysine-methyl-ester-hydrochloride/1431?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-z-d-lysine-methyl-ester-hydrochloride/1431?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart