CH6371377486 Chem Impex International, Inc.

Na,d-Bis-Z-L-ornithine

  • MDL number MFCD00037821
  • Cas Number : 2274-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-113� C
Synonym Z-L-Orn(Z)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-d-sup-i-i-bis-i-i-z-l-ornithine/1439?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-d-sup-i-i-bis-i-i-z-l-ornithine/1439?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart