CH6371377490 Chem Impex International, Inc.

Z-L-phenylalanine amide

  • MDL number MFCD00077028
  • Cas Number : 4801-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158 - 160 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-amide/1443?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-amide/1443?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart