CH6371377492 Chem Impex International, Inc.

Z-L-phenylalanine 4-nitrophenyl ester

  • MDL number MFCD00038119
  • Cas Number : 2578-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitrophenyl-ester/1445?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-phenylalanine-4-nitrophenyl-ester/1445?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart