CH6371377494 Chem Impex International, Inc.

Z-Phe-Gly-OH

  • MDL number MFCD00037782
  • Cas Number : 13122-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-152� C
Synonym Z-L-phenylalanyl-L-glycine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-phe-gly-oh/1447?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-phe-gly-oh/1447?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart