CH6371377499 Chem Impex International, Inc.

Z-Pro-Gly-OH

  • MDL number MFCD00077034
  • Cas Number : 2766-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-L-prolyl-L-glycine
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-pro-gly-oh/1454?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-pro-gly-oh/1454?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart