CH6371377500 Chem Impex International, Inc.

Z-Pro-Pro-OH

  • MDL number MFCD00077035
  • Cas Number : 7360-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-pro-pro-oh/1455?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-pro-pro-oh/1455?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart