CH6371377503 Chem Impex International, Inc.

Z-L-pyroglutamic acid 4-nitrophenyl ester

  • MDL number MFCD00077037
  • Cas Number : 40356-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 144 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Pale yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-pyroglutamic-acid-4-nitrophenyl-ester/1458?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-pyroglutamic-acid-4-nitrophenyl-ester/1458?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart